Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286134.1 | NP_001273063.1 | 5 | 2251 | ||
NM_021932.5 | NP_068751.4 | 5 | 2251 | |||
XM_005253052.3 | XP_005253109.1 | 5 | 1437 | |||
XM_017018107.1 | XP_016873596.1 | 5 | 1521 | |||
GenBank mRNA | AK022870.1 | - | 4 | 805 | ||
AK026897.1 | - | 5 | 2250 | |||
AK027090.1 | - | 2 | 178 | |||
AK027461.1 | - | 5 | 939 | |||
AK225420.1 | - | 5 | 937 | |||
AK314568.1 | - | 5 | 958 | |||
AL136935.1 | - | 5 | 1232 | |||
AL390088.1 | - | 5 | 972 | |||
BC011821.1 | - | 3 | 715 | |||
BC111499.1 | - | 5 | 1065 | 6526180 | ||
BC121807.1 | - | 5 | 1145 | 40108083 | ||
BC121808.1 | - | 5 | 1145 | 40108080 | ||
BX538115.1 | - | 3 | 1113 |