Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098673.1 | NP_001092143.1 | 3 | 416 | ||
NM_021934.4 | NP_068753.2 | 3 | 587 | |||
GenBank mRNA | AF218031.1 | - | 3 | 528 | ||
AK021835.1 | - | 3 | 585 | |||
BC005151.2 | - | 3 | 539 | 3508173 | ||
BC009937.2 | - | 3 | 521 | 4303452 | ||
BP309508.1 | - | 3 | 387 |