Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020318.2 | NP_064714.2 | 2 | 1608 | ||
NM_021936.2 | NP_068755.2 | 2 | 1608 | |||
XM_005245422.3 | XP_005245479.1 | 2 | 1655 | |||
XM_011509857.1 | XP_011508159.1 | 3 | 1525 | |||
XM_011509858.2 | XP_011508160.1 | 2 | 1868 | |||
XM_017002023.1 | XP_016857512.1 | 3 | 1706 | |||
XM_017002024.1 | XP_016857513.1 | 3 | 1753 | |||
XM_017002025.1 | XP_016857514.1 | 2 | 1859 | |||
GenBank mRNA | AF311940.1 | - | 1 | 444 | ||
AF342989.1 | - | 2 | 1608 | |||
AF342990.1 | - | 2 | 1608 | |||
AJ278348.1 | - | 1 | 407 | |||
BC117193.1 | - | 1 | 1216 | |||
BC117195.1 | - | 1 | 1217 | |||
BC152552.1 | - | 2 | 1596 |