Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 7 | 891 | ||
XM_005271642.2 | XP_005271699.1 | 6 | 780 | |||
XM_005271643.2 | XP_005271700.1 | 4 | 597 | |||
XM_006718894.2 | XP_006718957.1 | 6 | 667 | |||
XM_011542944.2 | XP_011541246.1 | 7 | 976 | |||
XM_011542945.2 | XP_011541247.1 | 8 | 936 | |||
XM_011542946.1 | XP_011541248.1 | 7 | 859 | |||
XM_011542947.2 | XP_011541249.1 | 5 | 545 | |||
XM_017018110.1 | XP_016873599.1 | 7 | 3726 | |||
XM_017018111.1 | XP_016873600.1 | 5 | 586 | |||
XM_017018112.1 | XP_016873601.1 | 4 | 597 | |||
GenBank mRNA | AB065001.1 | - | 4 | 569 | ||
AJ417081.1 | - | 4 | 541 | |||
AK023136.1 | - | 4 | 569 | |||
AK295857.1 | - | 6 | 758 | |||
AK298999.1 | - | 6 | 660 | |||
AK316001.1 | - | 6 | 691 | |||
AK316268.1 | - | 5 | 679 | |||
AL512682.1 | - | 2 | 214 | |||
BX648615.1 | - | 5 | 668 |