Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083953.1 | NP_001077422.1 | 8 | 1064 | ||
NM_001271643.1 | NP_001258572.1 | 8 | 1064 | |||
NM_001271644.1 | NP_001258573.1 | 8 | 852 | |||
NM_022065.4 | NP_071348.3 | 8 | 852 | |||
NR_073394.1 | 8 | 852 | ||||
XM_006712061.3 | XP_006712124.1 | 9 | 899 | |||
XM_006712062.1 | XP_006712125.1 | 8 | 852 | |||
XM_006712063.1 | XP_006712126.1 | 8 | 852 | |||
XM_006712064.2 | XP_006712127.1 | 8 | 852 | |||
XM_006712065.1 | XP_006712128.1 | 8 | 852 | |||
XM_006712066.1 | XP_006712129.1 | 8 | 852 | |||
XM_006712067.1 | XP_006712130.1 | 8 | 852 | |||
XM_006712068.3 | XP_006712131.1 | 8 | 852 | |||
XM_011533015.2 | XP_011531317.1 | 8 | 852 | |||
XM_017004674.1 | XP_016860163.1 | 8 | 852 | |||
XM_017004675.1 | XP_016860164.1 | 8 | 815 | |||
GenBank mRNA | AB051554.2 | - | 8 | 1064 | ||
AK126004.1 | - | 8 | 767 | |||
AK290023.1 | - | 8 | 852 | |||
AL832141.1 | - | 8 | 845 | |||
AY149629.1 | - | 8 | 844 | |||
AY149633.1 | - | 8 | 844 | |||
BC037990.2 | - | 8 | 829 | |||
BX641038.1 | - | 9 | 861 | |||
BX648155.1 | - | 8 | 848 |