Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022070.4 | NP_071353.4 | 12 | 1858 | ||
XM_005257570.3 | XP_005257627.1 | 13 | 2046 | |||
XM_011525110.2 | XP_011523412.1 | 11 | 1745 | |||
XM_011525111.2 | XP_011523413.1 | 11 | 1707 | |||
XM_017024936.1 | XP_016880425.1 | 12 | 2119 | |||
XM_017024937.1 | XP_016880426.1 | 12 | 3795 | |||
XM_017024938.1 | XP_016880427.1 | 12 | 1851 | |||
XM_017024939.1 | XP_016880428.1 | 12 | 2122 | |||
GenBank mRNA | AF349752.1 | - | 13 | 1803 | ||
AK025740.1 | - | 5 | 925 | |||
AK026070.1 | - | 5 | 698 | |||
AK127708.1 | - | 12 | 1852 | |||
AK314655.1 | - | 12 | 1861 | |||
BC119771.1 | - | 12 | 1848 | 40108435 | ||
BC119813.1 | - | 12 | 1849 | 40108426 | ||
BX640819.1 | - | 12 | 1849 | |||
BX640831.1 | - | 12 | 1849 | |||
CR627418.1 | - | 12 | 1848 |