Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031713.3 | NP_001026883.1 | 3 | 739 | ||
XM_011514802.1 | XP_011513104.1 | 3 | 742 | |||
GenBank mRNA | AK024611.1 | - | 4 | 1334 | ||
BC008234.1 | - | 3 | 494 | |||
BC016850.1 | - | 3 | 713 | 3886144 |