Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033053.2 | NP_149042.2 | 6 | 1064 | ||
XM_005262287.3 | XP_005262344.1 | 7 | 1143 | |||
XM_005262288.3 | XP_005262345.1 | 7 | 1168 | |||
XM_017029725.1 | XP_016885214.1 | 7 | 1174 | |||
GenBank mRNA | BC029799.1 | - | 6 | 1488 | ||
BC047596.1 | - | 5 | 1012 | 4830694 | ||
BI755761.1 | - | 1 | 160 |
Gene Symbol: DMRTC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080851.1 | NP_001074320.1 | 6 | 1064 | ||
XM_006724684.2 | XP_006724747.1 | 7 | 1143 | |||
XM_006724685.3 | XP_006724748.1 | 7 | 1168 | |||
XM_017029776.1 | XP_016885265.1 | 7 | 1174 |