Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190481.1 | NP_001177410.1 | 3 | 451 | ||
NM_022111.3 | NP_071394.2 | 3 | 451 | |||
XM_011541938.2 | XP_011540240.1 | 3 | 474 | |||
XM_011541939.2 | XP_011540241.1 | 3 | 468 | |||
XM_011541940.2 | XP_011540242.1 | 3 | 468 | |||
XM_011541941.2 | XP_011540243.1 | 3 | 468 | |||
GenBank mRNA | AF297866.1 | - | 3 | 451 | ||
AK308193.1 | - | 3 | 479 | |||
BC018670.1 | - | 3 | 471 | |||
BC038991.2 | - | 3 | 477 | |||
BC113116.1 | - | 3 | 444 | 7961377 | ||
BC115025.1 | - | 3 | 444 | 7961363 | ||
BC115026.1 | - | 3 | 444 | 7961366 | ||
BC137279.1 | - | 3 | 430 | 9021276 | ||
BC140789.1 | - | 3 | 430 | 9021657 |