Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIGNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042762.2 | NP_001036227.1 | 4 | 1207 | ||
NM_001287492.1 | NP_001274421.1 | 4 | 1645 | |||
NM_001287493.1 | NP_001274422.1 | 3 | 1039 | |||
NM_001287494.1 | NP_001274423.1 | 3 | 1034 | |||
NM_001287495.1 | NP_001274424.1 | 2 | 930 | |||
NM_001287496.1 | NP_001274425.1 | 4 | 924 | |||
NM_022116.4 | NP_071399.2 | 4 | 1200 | |||
XM_011515470.2 | XP_011513772.1 | 4 | 1209 | |||
XM_017012501.1 | XP_016867990.1 | 4 | 2556 | |||
XM_017012502.1 | XP_016867991.1 | 5 | 1728 | |||
GenBank mRNA | AK023142.1 | - | 4 | 1160 | ||
AK027537.1 | - | 4 | 1607 | |||
AK308136.1 | - | 4 | 801 | |||
AL834387.1 | - | 3 | 741 | |||
BC051867.1 | - | 4 | 1141 | 6158883 |