Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022117.3 | NP_071400.1 | 6 | 1686 | ||
XM_006724592.3 | XP_006724655.1 | 6 | 1719 | |||
XM_017029726.1 | XP_016885215.1 | 5 | 1851 | |||
XM_017029727.1 | XP_016885216.1 | 5 | 1851 | |||
GenBank mRNA | AB015345.1 | - | 6 | 926 | ||
AB209182.1 | - | 2 | 1578 | |||
AF254794.1 | - | 6 | 1425 | |||
AF273046.1 | - | 6 | 1676 | |||
AK131220.1 | - | 4 | 2133 | |||
AK292462.1 | - | 6 | 1679 | |||
AL713652.1 | - | 6 | 1665 | |||
AY040871.1 | - | 6 | 1675 | |||
BC001566.1 | - | 3 | 483 | |||
BC024270.1 | - | 6 | 1666 | 2819684 |