Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022120.1 | NP_071403.1 | 1 | 920 | ||
GenBank mRNA | AB050193.2 | - | 1 | 846 | ||
AK024440.1 | - | 1 | 25 | |||
AK093089.1 | - | 1 | 894 | |||
AK314586.1 | - | 1 | 827 | |||
AY013700.1 | - | 1 | 920 | |||
BC030015.1 | - | 1 | 839 |
Gene Symbol: OXCT2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132962.1 | 1 | 898 | |||
GenBank mRNA | BC072388.1 | - | 1 | 898 | 5588771 |