Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022151.4 | NP_071434.2 | 3 | 1294 | ||
GenBank mRNA | AF305550.1 | - | 1 | 924 | ||
AK024029.1 | - | 3 | 1266 | |||
AK315522.1 | - | 3 | 1306 | |||
BC015044.1 | - | 3 | 1228 | 3922055 |