Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022159.3 | NP_071442.2 | 10 | 1603 | ||
GenBank mRNA | AB032017.1 | - | 10 | 1564 | ||
AF192403.1 | - | 8 | 1264 | |||
AK129766.1 | - | 1 | 55 | |||
AK172750.1 | - | 7 | 1082 | |||
AK292894.1 | - | 10 | 1496 | |||
AK313738.1 | - | 10 | 1602 | |||
AY358360.1 | - | 10 | 1470 | |||
BC025721.1 | - | 10 | 1469 | 5229055 |
Set Membership: |
G-protein coupled receptor |