Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022353.2 | NP_071748.2 | 2 | 669 | ||
XM_005246766.3 | XP_005246823.1 | 2 | 269 | |||
XM_006712685.1 | XP_006712748.1 | 2 | 381 | |||
XM_006712686.1 | XP_006712749.1 | 2 | 257 | |||
XM_011511631.1 | XP_011509933.1 | 2 | 400 | |||
XM_017004676.1 | XP_016860165.1 | 2 | 269 | |||
XM_017004677.1 | XP_016860166.1 | 2 | 269 | |||
XM_017004678.1 | XP_016860167.1 | 2 | 381 | |||
XM_017004679.1 | XP_016860168.1 | 2 | 400 | |||
XM_017004680.1 | XP_016860169.1 | 2 | 355 | |||
XM_017004682.1 | XP_016860171.1 | 2 | 257 | |||
XM_017004683.1 | XP_016860172.1 | 2 | 257 | |||
XM_017004685.1 | XP_016860174.1 | 2 | 395 | |||
GenBank mRNA | AJ295148.1 | - | 1 | 158 | ||
AK055441.1 | - | 3 | 478 | |||
AK294841.1 | - | 2 | 266 | |||
AK314565.1 | - | 2 | 669 | |||
BC011904.2 | - | 2 | 238 | 4121450 |