Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2BPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024496.3 | NP_078772.1 | 1 | 2162 | ||
GenBank mRNA | AB058768.1 | - | 2 | 1657 | ||
BC152431.1 | - | 2 | 1657 |