Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023945.2 | NP_076434.2 | 4 | 506 | ||
GenBank mRNA | AB013103.1 | - | 4 | 471 | ||
AF237907.1 | - | 4 | 474 | |||
AF321127.1 | - | 4 | 476 | |||
BC029884.1 | - | 4 | 506 | 5171526 |