Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005474.2 | NP_001005474.1 | 12 | 2035 | ||
NM_031419.3 | NP_113607.1 | 11 | 2168 | |||
GenBank mRNA | AB037925.1 | - | 11 | 2101 | ||
AF548362.1 | - | 12 | 2032 | |||
AK054787.1 | - | 12 | 1904 | |||
AK091782.1 | - | 12 | 1802 | |||
BC014587.2 | - | 2084 | ||||
BC060800.1 | - | 11 | 2168 | 30344780 | ||
DQ224339.1 | - | 11 | 1957 |