Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 4 | 437 | ||
NM_022463.4 | NP_071908.2 | 4 | 780 | |||
XM_005256756.4 | XP_005256813.1 | 4 | 786 | |||
XM_005256758.3 | XP_005256815.1 | 4 | 1109 | |||
XM_017024949.1 | XP_016880438.1 | 4 | 788 | |||
GenBank mRNA | AK022676.1 | - | 2 | 209 | ||
AK027451.1 | - | 3 | 333 | |||
AK297990.1 | - | 1 | 89 | |||
AK302073.1 | - | 4 | 439 | |||
AW752727.1 | - | 4 | 369 | |||
AY189690.1 | - | 3 | 320 | |||
BC009327.2 | - | 4 | 374 | |||
BC063828.1 | - | 1 | 35 | |||
BC073845.1 | - | 2 | 190 | |||
BC104634.1 | - | 4 | 473 | |||
CX786695.1 | - | 4 | 780 |