Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022483.4 | NP_071928.2 | 2 | 411 | ||
XM_005248333.4 | XP_005248390.1 | 3 | 1279 | |||
XM_005248335.3 | XP_005248392.1 | 3 | 563 | |||
XM_006714486.3 | XP_006714549.1 | 2 | 883 | |||
XM_011514074.2 | XP_011512376.1 | 4 | 1655 | |||
XM_011514075.2 | XP_011512377.1 | 4 | 1651 | |||
GenBank mRNA | AK315466.1 | - | 3 | 563 | ||
BC013351.2 | - | 3 | 600 | 3621124 | ||
BC063832.1 | - | 1 | 357 | 6178186 | ||
BC119658.1 | - | 1 | 317 | 40115985 | ||
BC119659.1 | - | 1 | 317 | 40115986 | ||
DA028175.1 | - | 2 | 412 | |||
DA035926.1 | - | 2 | 380 |