Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077525.2 | NP_001070993.1 | 2 | 368 | ||
NM_001077526.2 | NP_001070994.1 | 2 | 368 | |||
NM_022485.4 | NP_071930.2 | 2 | 368 | |||
XM_005265400.1 | XP_005265457.1 | 2 | 341 | |||
XM_017007034.1 | XP_016862523.1 | 3 | 538 | |||
XM_017007035.1 | XP_016862524.1 | 3 | 538 | |||
XM_017007036.1 | XP_016862525.1 | 3 | 537 | |||
XM_017007037.1 | XP_016862526.1 | 3 | 538 | |||
XM_017007038.1 | XP_016862527.1 | 2 | 341 | |||
XM_017007039.1 | XP_016862528.1 | 3 | 537 | |||
XM_017007040.1 | XP_016862529.1 | 3 | 538 | |||
XM_017007042.1 | XP_016862531.1 | 2 | 341 | |||
XM_017007044.1 | XP_016862533.1 | 2 | 335 | |||
XM_017007045.1 | XP_016862534.1 | 2 | 310 | |||
GenBank mRNA | AF289603.1 | - | 2 | 331 | ||
AK074792.1 | - | 2 | 339 | |||
AK295891.1 | - | 2 | 296 | |||
AK298846.1 | - | 2 | 289 | |||
AL713695.1 | - | 2 | 289 | |||
BC001674.1 | - | 2 | 273 | 2961594 | ||
BC025952.1 | - | 2 | 318 | 4876556 | ||
BC035690.1 | - | 2 | 280 | 5755887 | ||
BC050626.1 | - | 2 | 291 | 4416416 | ||
DA604126.1 | - | 2 | 341 | |||
DC316206.1 | - | 2 | 370 | |||
DQ630520.1 | - | 2 | 217 |