Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253775.1 | NP_001240704.1 | 2 | 560 | ||
NM_001318246.1 | NP_001305175.1 | 2 | 147 | |||
NM_194071.3 | NP_919047.2 | 2 | 560 | |||
XM_005250541.4 | XP_005250598.1 | 2 | 939 | |||
XM_017012540.1 | XP_016868029.1 | 3 | 1962 | |||
XM_017012541.1 | XP_016868030.1 | 3 | 525 | |||
GenBank mRNA | AJ549092.1 | - | 2 | 516 | ||
AJ549387.1 | - | 2 | 516 | |||
AK131517.1 | - | 2 | 526 | |||
AK293048.1 | - | 2 | 492 | |||
BC046181.1 | - | 1 | 24 | |||
BC063666.1 | - | 2 | 506 | |||
BC110813.1 | - | 2 | 499 | 3923538 | ||
BC131723.1 | - | 2 | 325 | |||
BQ711676.1 | - | 2 | 560 | |||
BX649143.1 | - | 2 | 155 | |||
CN394705.1 | - | 2 | 561 | |||
CR749563.1 | - | 2 | 287 |