Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288794.1 | NP_001275723.1 | 5 | 598 | ||
NM_001288795.1 | NP_001275724.1 | 4 | 447 | |||
NM_022762.4 | NP_073599.2 | 4 | 458 | |||
XM_005265969.3 | XP_005266026.1 | 3 | 278 | |||
XM_005265971.4 | XP_005266028.1 | 4 | 1025 | |||
XM_005265972.3 | XP_005266029.1 | 3 | 405 | |||
XM_017009727.1 | XP_016865216.1 | 3 | 1069 | |||
XM_017009728.1 | XP_016865217.1 | 3 | 736 | |||
GenBank mRNA | AK093481.1 | - | 4 | 399 | ||
AK094065.1 | - | 3 | 268 | |||
AK122692.1 | - | 1 | 984 | |||
AK125337.1 | - | 5 | 545 | |||
AL713670.1 | - | 3 | 321 | |||
AY359092.1 | - | 1 | 1316 | |||
BC009911.2 | - | 4 | 392 | 2820100 | ||
DB053605.1 | - | 4 | 458 | |||
DQ494789.1 | - | 4 | 392 |