Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTHFSD SIRNA Assays ›
Gene Symbol:
MTHFSD
Gene Name:
methenyltetrahydrofolate synthetase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.16: 86530176 - 86555235 on Build GRCh38
UniGene ID:
Hs.343627
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34925
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTHFSD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159377.1
NP_001152849.1
4
333
NM_001159378.1
NP_001152850.1
4
333
NM_001159379.1
NP_001152851.1
4
330
NM_001159380.1
NP_001152852.1
4
297
NM_022764.2
NP_073601.2
4
330
NR_027489.1
4
326
NR_027490.1
4
393
XM_005256101.1
XP_005256158.1
4
373
XM_005256105.1
XP_005256162.1
4
379
XM_005256106.1
XP_005256163.1
4
288
XM_006721247.3
XP_006721310.1
3
204
XM_011523280.2
XP_011521582.1
4
373
XM_011523282.2
XP_011521584.1
4
294
XM_011523283.2
XP_011521585.1
4
294
XM_011523284.2
XP_011521586.1
4
333
XM_011523285.1
XP_011521587.1
4
294
XM_011523286.1
XP_011521588.1
4
394
XM_011523287.1
XP_011521589.1
4
298
XM_011523288.1
XP_011521590.1
3
226
XM_011523289.1
XP_011521591.1
4
394
XM_017023571.1
XP_016879060.1
3
219
XM_017023572.1
XP_016879061.1
3
216
XM_017023573.1
XP_016879062.1
4
287
XM_017023574.1
XP_016879063.1
3
219
GenBank mRNA
AK023060.1
-
4
308
AK023955.1
-
4
287
AK096836.1
-
4
326
AK297726.1
-
4
330
BC013911.2
-
3
264
BC112057.1
-
4
313
8327525
BC112059.1
-
4
313
8327527
BC143702.1
-
4
298
9052216
BC143704.1
-
4
298
BC143705.1
-
4
297
BC143707.1
-
4
301
9052221
Back To Top
More Information
Panther Classification:
Molecular Function -
ligase
Gene Ontology Categories:
Process(es)
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0