Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022780.3 | NP_073617.1 | 7, 8 | 1327 | ||
GenBank mRNA | AK022815.1 | - | 7, 8 | 1304 | ||
AK025448.1 | - | 3, 4 | 408 | |||
AK303113.1 | - | 3, 4 | 353 | |||
AL832022.1 | - | 2, 3 | 243 | |||
BC012165.1 | - | 7, 8 | 1048 | 4636136 | ||
BC047668.1 | - | 7, 8 | 1257 | 6066798 |