Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IQCH SIRNA Assays ›
Gene Symbol:
IQCH
Gene Name:
IQ motif containing H
Gene Aliases:
NYDSP5
Chromosome Location:
Chr.15: 67254772 - 67501804 on Build GRCh38
UniGene ID:
Hs.657894
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34973
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IQCH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031715.2
NP_001026885.1
9
1035
NM_001284347.1
NP_001271276.1
5
572
NM_001284348.1
NP_001271277.1
7
807
NM_001284349.1
NP_001271278.1
6
701
NM_001322470.1
NP_001309399.1
6
701
NM_001322471.1
NP_001309400.1
5
572
NM_001322472.1
NP_001309401.1
5
572
NM_001322473.1
NP_001309402.1
6
701
NM_001322474.1
NP_001309403.1
6
701
NM_001322475.1
NP_001309404.1
7
819
XM_011521907.2
XP_011520209.1
10
1176
XM_011521908.2
XP_011520210.1
9
1035
XM_011521911.2
XP_011520213.1
8
963
XM_011521912.2
XP_011520214.1
10
1176
XM_011521913.2
XP_011520215.1
10
1177
XM_011521914.2
XP_011520216.1
7
695
XM_011521915.2
XP_011520217.1
10
1176
XM_011521916.2
XP_011520218.1
6
614
XM_011521917.2
XP_011520219.1
8
883
XM_011521918.2
XP_011520220.1
6
669
XM_017022491.1
XP_016877980.1
10
1176
XM_017022492.1
XP_016877981.1
10
1176
XM_017022493.1
XP_016877982.1
9
1137
XM_017022494.1
XP_016877983.1
10
1176
XM_017022495.1
XP_016877984.1
10
1176
XM_017022496.1
XP_016877985.1
10
1176
XM_017022497.1
XP_016877986.1
10
1176
XM_017022498.1
XP_016877987.1
10
1176
XM_017022499.1
XP_016877988.1
10
1176
XM_017022500.1
XP_016877989.1
7
842
XM_017022501.1
XP_016877990.1
6
669
XM_017022502.1
XP_016877991.1
7
830
XM_017022503.1
XP_016877992.1
5
595
XM_017022504.1
XP_016877993.1
5
595
GenBank mRNA
AK022538.1
-
7
757
AK292478.1
-
9
987
AY014282.2
-
9
1006
BC031041.1
-
6
653
BC039292.1
-
5
522
5270043
BC048964.1
-
9
985
5297681
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0