Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134774.1 | NP_001128246.1 | 4, 5 | 700 | ||
NM_001134775.1 | NP_001128247.1 | 5, 6 | 994 | |||
NM_001134776.1 | NP_001128248.1 | 5, 6 | 873 | |||
NM_001318734.1 | NP_001305663.1 | 5, 6 | 931 | |||
NM_022822.2 | NP_073733.1 | 5, 6 | 968 | |||
XM_005274183.1 | XP_005274240.1 | 5, 6 | 910 | |||
XM_005274184.1 | XP_005274241.1 | 5, 6 | 931 | |||
GenBank mRNA | AK022449.1 | - | 5, 6 | 860 | ||
AK022907.1 | - | 5, 6 | 913 | |||
AK094593.1 | - | 3, 4 | 570 | |||
AK126489.1 | - | 3, 4 | 1352 | |||
AK315725.1 | - | 5, 6 | 863 | |||
AL136864.1 | - | 5, 6 | 894 | |||
BC034373.1 | - | 5, 6 | 856 | 4342879 | ||
BE882786.1 | - | 4, 5 | 595 |