Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PORCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282167.1 | NP_001269096.1 | 7 | 1063 | ||
NM_022825.3 | NP_073736.2 | 7 | 918 | |||
NM_203473.2 | NP_982299.1 | 8 | 936 | |||
NM_203474.1 | NP_982300.1 | 7 | 788 | |||
NM_203475.2 | NP_982301.1 | 9 | 951 | |||
XM_005272635.2 | XP_005272692.1 | 8 | 1291 | |||
XM_005272636.2 | XP_005272693.1 | 7 | 1273 | |||
XM_005272637.1 | XP_005272694.1 | 10 | 1233 | |||
XM_006724545.3 | XP_006724608.1 | 10 | 1016 | |||
XM_006724546.3 | XP_006724609.1 | 10 | 1088 | |||
XM_006724547.1 | XP_006724610.1 | 9 | 808 | |||
XM_011543948.2 | XP_011542250.1 | 7 | 1202 | |||
XM_017029735.1 | XP_016885224.1 | 9 | 1316 | |||
XM_017029736.1 | XP_016885225.1 | 7 | 775 | |||
GenBank mRNA | AF317058.1 | - | 8 | 772 | ||
AF317059.1 | - | 7 | 757 | |||
AF317060.1 | - | 7 | 754 | |||
AF317061.1 | - | 6 | 739 | |||
AK056340.1 | - | 7 | 1245 | |||
AK057727.1 | - | 6 | 1165 | |||
AK293347.1 | - | 7 | 1028 | |||
AK314745.1 | - | 8 | 821 | |||
AK316378.1 | - | 6 | 810 | |||
AL579208.3 | - | 7 | 837 | |||
BC019080.2 | - | 8 | 883 | 4559533 | ||
L08239.1 | - | 7 | 688 |