Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 12 | 1702 | ||
NM_001258373.1 | NP_001245302.1 | 13 | 1805 | |||
NM_022827.3 | NP_073738.2 | 13 | 1750 | |||
GenBank mRNA | AK025000.1 | - | 12 | 1599 | ||
AK025622.1 | - | 8 | 991 | |||
AK092731.1 | - | 13 | 1735 | |||
AK125807.1 | - | 14 | 2572 | |||
AK225189.1 | - | 12 | 1599 | |||
AK291805.1 | - | 13 | 1693 | |||
BC017468.1 | - | 2 | 194 | |||
BC025255.1 | - | 13 | 1638 | 5019010 | ||
BC065526.1 | - | 13 | 1692 | 6149633 | ||
BC111029.1 | - | 12 | 1607 | 5245538 | ||
CR936769.1 | - | 11 | 1940 | |||
DQ238597.1 | - | 12 | 1700 | |||
GQ472220.1 | - | 12 | 1607 |