Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022828.3 | NP_073739.3 | 27 | 4072 | ||
XM_011543581.1 | XP_011541883.1 | 27 | 4083 | |||
XM_011543582.2 | XP_011541884.1 | 27 | 4057 | |||
XM_017009730.1 | XP_016865219.1 | 27 | 4083 | |||
XM_017009731.1 | XP_016865220.1 | 26 | 3858 | |||
XM_017009732.1 | XP_016865221.1 | 26 | 3858 | |||
XM_017009733.1 | XP_016865222.1 | 25 | 3686 | |||
XM_017009734.1 | XP_016865223.1 | 25 | 3686 | |||
XM_017009735.1 | XP_016865224.1 | 24 | 3505 | |||
GenBank mRNA | AK025593.1 | - | 11 | 1685 | ||
AK131450.1 | - | 26 | 3765 | |||
AK225316.1 | - | 11 | 1685 | |||
BC137285.1 | - | 27 | 4022 | 9021282 | ||
CB268902.1 | - | 3 | 439 | |||
CR627037.1 | - | 2 | 3259 |