Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022841.5 | NP_073752.5 | 8 | 1021 | ||
XM_005254603.3 | XP_005254660.2 | 9 | 1115 | |||
XM_011521925.2 | XP_011520227.1 | 8 | 953 | |||
XM_017022506.1 | XP_016877995.1 | 9 | 3838 | |||
XM_017022507.1 | XP_016877996.1 | 8 | 1081 | |||
XM_017022508.1 | XP_016877997.1 | 7 | 2250 | |||
GenBank mRNA | AK126103.1 | - | 1 | 173 | ||
AK128045.1 | - | 8 | 958 | |||
BC112936.1 | - | 6 | 841 | 6014850 | ||
BC143438.1 | - | 6 | 778 |