Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286191.1 | NP_001273120.1 | 4, 5 | 483 | ||
NM_001286192.1 | NP_001273121.1 | 4, 5 | 390 | |||
NM_001286197.1 | NP_001273126.1 | 3, 4 | 291 | |||
NM_001286198.1 | NP_001273127.1 | 5, 6 | 647 | |||
NM_022895.2 | NP_075046.1 | 4, 5 | 390 | |||
NR_104409.1 | 4, 5 | 400 | ||||
XM_017019829.1 | XP_016875318.1 | 4, 5 | 474 | |||
GenBank mRNA | AK022510.1 | - | 4, 5 | 385 | ||
AK222630.1 | - | 4, 5 | 375 | |||
AK225162.1 | - | 4, 5 | 385 | |||
AK299170.1 | - | 5, 6 | 633 | |||
AK307971.1 | - | 4, 5 | 485 | |||
BC014661.1 | - | 4, 5 | 364 | 4860511 | ||
BP201132.1 | - | 4, 5 | 370 | |||
BP343829.1 | - | 3, 4 | 218 | |||
DA014862.1 | - | 4, 5 | 389 |