Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318767.1 | NP_001305696.1 | 3 | 245 | ||
NM_001318768.1 | NP_001305697.1 | 3 | 189 | |||
NM_001318769.1 | NP_001305698.1 | 3 | 178 | |||
NM_001318770.1 | NP_001305699.1 | 3 | 451 | |||
NM_001318771.1 | NP_001305700.1 | 2 | 195 | |||
NM_032111.3 | NP_115487.2 | 3 | 284 | |||
GenBank mRNA | AK312086.1 | - | 3 | 216 | ||
BC065005.1 | - | 2 | 140 | 6052682 | ||
BG545603.1 | - | 3 | 449 | |||
BM013002.1 | - | 2 | 110 | |||
BM559405.1 | - | 296 | ||||
BM910935.1 | - | 247 | ||||
BU597168.1 | - | 3 | 211 | |||
CK823336.1 | - | 3 | 189 |