Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261435.1 | NP_001248364.1 | 7 | 766 | ||
NM_001261436.1 | NP_001248365.1 | 7 | 766 | |||
NM_139158.2 | NP_631897.1 | 7 | 676 | |||
XM_005246781.3 | XP_005246838.1 | 7 | 766 | |||
XM_005246782.4 | XP_005246839.1 | 8 | 1013 | |||
XM_011511650.2 | XP_011509952.1 | 7 | 766 | |||
XM_011511652.1 | XP_011509954.1 | 7 | 766 | |||
XM_011511654.2 | XP_011509956.1 | 7 | 766 | |||
XM_011511655.2 | XP_011509957.1 | 4 | 773 | |||
XM_011511656.2 | XP_011509958.1 | 4 | 380 | |||
XM_011511657.2 | XP_011509959.1 | 2 | 277 | |||
GenBank mRNA | AB053308.1 | - | 7 | 630 | ||
AK131512.1 | - | 6 | 703 | |||
AK292434.1 | - | 8 | 883 | |||
AK310050.1 | - | 9 | 999 | |||
AX772791.1 | - | 7 | 630 | |||
BC038807.1 | - | 7 | 724 | 5200755 | ||
BX647394.1 | - | 7 | 730 |