Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023008.3 | NP_075384.3 | 4 | 370 | ||
XM_011528190.2 | XP_011526492.1 | 3 | 301 | |||
GenBank mRNA | AK023011.1 | - | 2 | 148 | ||
AK093879.1 | - | 3 | 482 | |||
AK300780.1 | - | 4 | 354 | |||
AK300850.1 | - | 4 | 375 | |||
AL833189.1 | - | 3 | 1023 | |||
AU280109.1 | - | 4 | 371 | |||
BC002890.2 | - | 1 | 35 | |||
BC112249.1 | - | 1 | 27 | |||
BC112251.1 | - | 1 | 27 | |||
BC143770.1 | - | 1 | 27 | |||
DA438802.1 | - | 4 | 371 |