Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023008.3 | NP_075384.3 | 12 | 1131 | ||
XM_011528190.2 | XP_011526492.1 | 11 | 1062 | |||
GenBank mRNA | AK023011.1 | - | 10 | 909 | ||
AK093879.1 | - | 11 | 1243 | |||
AK097181.1 | - | 3 | 449 | |||
AK300780.1 | - | 12 | 1115 | |||
AK300850.1 | - | 12 | 1136 | |||
AL833189.1 | - | 11 | 1784 | |||
BC002890.2 | - | 9 | 796 | |||
BC009876.2 | - | 3 | 303 | |||
BC112249.1 | - | 9 | 788 | |||
BC112251.1 | - | 9 | 788 | |||
BC143770.1 | - | 9 | 788 |