Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MPPE1 SIRNA Assays ›
Gene Symbol:
MPPE1
Gene Name:
metallophosphoesterase 1
Gene Aliases:
PGAP5
Chromosome Location:
Chr.18: 11883471 - 11908797 on Build GRCh38
UniGene ID:
Hs.712666
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35258
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MPPE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242904.1
NP_001229833.1
6
1311
NM_001319154.1
NP_001306083.1
7
1418
NM_023075.5
NP_075563.3
7
1418
NR_040241.1
7
1418
NR_040242.3
8
1286
NR_040243.1
7
1418
XM_006722340.3
XP_006722403.1
7
971
XM_006722341.3
XP_006722404.1
9
1593
XM_006722342.3
XP_006722405.1
8
1377
XM_006722343.3
XP_006722406.1
8
1498
XM_006722345.3
XP_006722408.1
6
1311
XM_006722346.2
XP_006722409.1
6
1311
XM_006722347.3
XP_006722410.1
7
971
XM_006722348.2
XP_006722411.1
7
971
XM_011525731.2
XP_011524033.1
10
2001
XM_011525732.1
XP_011524034.1
7
971
XM_011525734.1
XP_011524036.1
7
971
XM_017025920.1
XP_016881409.1
9
1906
XM_017025921.1
XP_016881410.1
6
813
XM_017025922.1
XP_016881411.1
9
1593
XM_017025923.1
XP_016881412.1
10
2001
XM_017025924.1
XP_016881413.1
8
1377
XM_017025925.1
XP_016881414.1
8
1498
XM_017025926.1
XP_016881415.1
9
1906
XM_017025927.1
XP_016881416.1
8
1377
XM_017025928.1
XP_016881417.1
8
1498
XM_017025929.1
XP_016881418.1
6
1311
XM_017025930.1
XP_016881419.1
6
1311
XM_017025931.1
XP_016881420.1
7
971
GenBank mRNA
AF289560.1
-
7
939
AF363484.1
-
7
1420
AK021647.1
-
2
71
AK023052.1
-
5
634
AK054639.1
-
8
784
AK223450.1
-
6
791
AK309948.1
-
7
936
AL833636.1
-
7
970
AX775853.1
-
7
968
BC002877.2
-
6
798
3942160
BC073994.1
-
7
930
6180123
Back To Top
More Information
Panther Classification:
Molecular Function -
esterase
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
ER to Golgi vesicle-mediated transport
protein binding
phosphoric diester hydrolase activity
manganese ion binding
GPI anchor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0