Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023077.2 | NP_075565.2 | 3 | 559 | ||
GenBank mRNA | AK022501.1 | - | 3 | 559 | ||
AK225041.1 | - | 3 | 534 | |||
AK225087.1 | - | 3 | 540 | |||
BC015313.1 | - | 3 | 525 | 4430419 |