Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321299.1 | NP_001308228.1 | 2 | 771 | ||
NM_032387.4 | NP_115763.2 | 2 | 771 | |||
XM_011525135.2 | XP_011523437.1 | 2 | 1701 | |||
XM_017024962.1 | XP_016880451.1 | 2 | 1701 | |||
XM_017024963.1 | XP_016880452.1 | 2 | 1701 | |||
XM_017024964.1 | XP_016880453.1 | 2 | 1701 | |||
XM_017024965.1 | XP_016880454.1 | 2 | 1701 | |||
GenBank mRNA | AF390018.1 | - | 2 | 724 | ||
AJ309861.1 | - | 1 | 85 | |||
AJ316534.1 | - | 2 | 720 | |||
BC136664.1 | - | 2 | 721 | 9020654 |