Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35286
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
14
1531
NM_001289059.1
NP_001275988.1
11
1238
NM_001289061.1
NP_001275990.1
13
1796
NM_030906.3
NP_112168.1
14
1945
XM_011520288.2
XP_011518590.1
14
3023
XM_011520290.2
XP_011518592.1
16
1942
XM_011520292.2
XP_011518594.1
14
1744
XM_011520293.2
XP_011518595.1
14
1773
XM_011520294.1
XP_011518596.1
13
1738
XM_011520295.2
XP_011518597.1
13
3834
XM_011520297.2
XP_011518599.1
13
1511
XM_017018143.1
XP_016873632.1
16
2002
XM_017018144.1
XP_016873633.1
16
1904
XM_017018145.1
XP_016873634.1
15
3936
XM_017018146.1
XP_016873635.1
16
2066
XM_017018147.1
XP_016873636.1
15
3863
XM_017018148.1
XP_016873637.1
14
3795
XM_017018149.1
XP_016873638.1
15
1947
XM_017018150.1
XP_016873639.1
15
1802
XM_017018151.1
XP_016873640.1
15
1761
XM_017018152.1
XP_016873641.1
13
2092
XM_017018153.1
XP_016873642.1
13
1690
XM_017018154.1
XP_016873643.1
13
1549
XM_017018155.1
XP_016873644.1
13
2104
XM_017018156.1
XP_016873645.1
13
1550
XM_017018157.1
XP_016873646.1
13
1315
XM_017018158.1
XP_016873647.1
14
1601
XM_017018159.1
XP_016873648.1
14
3741
XM_017018160.1
XP_016873649.1
12
1540
XM_017018165.1
XP_016873654.1
14
1504
GenBank mRNA
AJ303380.2
-
12
2289
AK092720.1
-
11
1177
AK093251.1
-
13
1654
AK122808.1
-
9
3666
AK293193.1
-
14
1397
AL833011.1
-
6
437
BC031231.1
-
14
1893
5296502
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0