Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019091.3 | NP_061964.3 | 3 | 691 | ||
XM_011511689.2 | XP_011509991.1 | 4 | 800 | |||
GenBank mRNA | AF286162.1 | - | 3 | 530 | ||
AK000074.1 | - | 1 | 94 | |||
AK074209.1 | - | 3 | 666 | |||
AK314459.1 | - | 3 | 695 | |||
BC044567.1 | - | 3 | 385 | 5262097 |