Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023934.3 | NP_076423.2 | 2 | 382 | ||
GenBank mRNA | AF267862.1 | - | 1 | 412 | ||
AF320778.1 | - | 2 | 345 | |||
AJ272054.1 | - | 2 | 339 | |||
AK313143.1 | - | 2 | 324 | |||
AY032594.1 | - | 2 | 232 | |||
BC000255.1 | - | 2 | 329 | 3355861 | ||
BC072685.1 | - | 2 | 287 | 6502640 | ||
BC108657.1 | - | 2 | 238 | 5752287 | ||
BI463291.1 | - | 2 | 387 |