Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 2 | 361 | ||
NM_001024939.3 | NP_001020110.1 | 2 | 249 | |||
NM_001282864.1 | NP_001269793.1 | 2 | 604 | |||
NM_030807.4 | NP_110434.3 | 3 | 472 | |||
NR_104247.1 | 2 | 361 | ||||
NR_104248.1 | 3 | 472 | ||||
GenBank mRNA | AF443201.1 | - | 3 | 303 | ||
AF479409.1 | - | 1 | 42 | |||
AJ271290.1 | - | 2 | 192 | |||
AK075467.1 | - | 2 | 137 | |||
AK296169.1 | - | 2 | 130 | |||
AK300196.1 | - | 2 | 604 | |||
AK304409.1 | - | 2 | 227 | |||
AK308589.1 | - | 2 | 227 | |||
AK308602.1 | - | 2 | 130 | |||
AK314502.1 | - | 3 | 243 | |||
AK315937.1 | - | 2 | 132 | |||
AK316049.1 | - | 2 | 198 | |||
BC063525.1 | - | 3 | 229 | |||
BC094735.1 | - | 2 | 178 | 5304025 | ||
BC100807.2 | - | 2 | 111 | |||
BC100808.2 | - | 2 | 107 | 40000757 | ||
BC100809.1 | - | 2 | 107 | 40000760 | ||
BC100810.1 | - | 2 | 107 | |||
CR456373.1 | - | 2 | 138 | |||
DA084206.1 | - | 2 | 361 | |||
DC303452.1 | - | 2 | 250 |