Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024091.3 | NP_076996.2 | 7 | 2109 | ||
NR_036553.1 | 6 | 558 | ||||
NR_073608.1 | 6 | 679 | ||||
XM_006714498.1 | XP_006714561.1 | 7 | 2020 | |||
GenBank mRNA | AF161354.1 | - | 6 | 532 | ||
AK026927.1 | - | 7 | 2074 | |||
BC001295.1 | - | 6 | 1601 | |||
BC113563.1 | - | 6 | 2024 | 8322615 | ||
BC143517.1 | - | 6 | 2145 | |||
BC143518.1 | - | 7 | 1461 |