Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024097.3 | NP_077002.2 | 3 | 305 | ||
NR_040733.1 | 2 | 189 | ||||
GenBank mRNA | BC001508.2 | - | 3 | 281 | 2959109 | |
BC001711.1 | - | 3 | 281 | 3543543 | ||
BM702424.1 | - | 2 | 141 | |||
CN256666.1 | - | 2 | 89 | |||
DA684965.1 | - | 3 | 305 |