Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142776.1 | NP_001136248.1 | 4 | 871 | ||
NM_024111.3 | NP_077016.2 | 3 | 1006 | |||
XM_017022563.1 | XP_016878052.1 | 4 | 5530 | |||
XM_017022564.1 | XP_016878053.1 | 5 | 3312 | |||
XM_017022565.1 | XP_016878054.1 | 4 | 5519 | |||
GenBank mRNA | BC001683.1 | - | 3 | 960 | 2964714 | |
BC001847.2 | - | 3 | 951 | 2964714 | ||
BC019625.1 | - | 3 | 962 | 4473135 | ||
BM784841.1 | - | 1 | 332 |