Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 4 | 535 | ||
NM_001011668.2 | NP_001011668.1 | 4 | 491 | |||
NM_001011669.2 | NP_001011669.1 | 4 | 515 | |||
NM_001011670.2 | NP_001011670.1 | 3 | 387 | |||
NM_001011671.2 | NP_001011671.1 | 3 | 343 | |||
NM_024300.4 | NP_077276.2 | 4 | 471 | |||
NR_133934.1 | 4 | 535 | ||||
NR_133936.1 | 4 | 491 | ||||
NR_133937.1 | 3 | 383 | ||||
GenBank mRNA | AK095922.1 | - | 3 | 192 | ||
AK098285.1 | - | 2 | 2994 | |||
AK225982.1 | - | 4 | 330 | |||
AK290088.1 | - | 4 | 320 | |||
AY070434.1 | - | 4 | 363 | |||
BC002546.2 | - | 4 | 310 | 3139026 | ||
BM695222.1 | - | 3 | 227 | |||
BX473986.1 | - | 4 | 373 | |||
DA367603.1 | - | 4 | 384 | |||
DA988698.1 | - | 4 | 340 |