Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250589.3 | XP_005250646.1 | 4, 5 | 663 | ||
XM_017012617.1 | XP_016868106.1 | 4, 5 | 786 | |||
XM_017012618.1 | XP_016868107.1 | 4, 5 | 530 | |||
GenBank mRNA | AK313794.1 | - | 3, 4 | 465 | ||
BC002837.2 | - | 3, 4 | 447 | 3634983 | ||
BP236768.1 | - | 4, 5 | 557 | |||
BT006973.1 | - | 3, 4 | 235 | |||
CB993361.1 | - | 4, 5 | 704 | |||
DB241717.1 | - | 3, 4 | 334 |