Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024323.3 | NP_077299.3 | 4 | 307 | ||
XM_006722885.3 | XP_006722948.2 | 4 | 680 | |||
XM_011528286.2 | XP_011526588.1 | 3 | 469 | |||
XM_011528287.2 | XP_011526589.1 | 3 | 129 | |||
XM_017027300.1 | XP_016882789.1 | 3 | 467 | |||
XM_017027301.1 | XP_016882790.1 | 3 | 331 | |||
GenBank mRNA | AK097431.1 | - | 4 | 307 | ||
BC119719.1 | - | 3 | 275 | 40117748 |