Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024333.2 | NP_077309.1 | 6 | 552 | ||
XM_005259644.2 | XP_005259701.1 | 6 | 551 | |||
GenBank mRNA | AF316829.1 | - | 6 | 443 | ||
AK021750.1 | - | 6 | 452 | |||
AK295523.1 | - | 6 | 447 | |||
AK303606.1 | - | 6 | 436 | |||
AK315661.1 | - | 6 | 546 | |||
AY032617.1 | - | 6 | 475 | |||
BC003124.1 | - | 6 | 452 | 3502614 |